Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 3-(3-amino-4-(methylamino)-N-(pyridin-2-yl)benzamido)propanoate3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester

E1033-1G E1033-5G
TCE1033-5GEA 92.32 USD
TCE1033-5G TCE1033-1G
Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate ≥98.0% (by HPLC, titration analysis)
Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate

CAS Number: 212322-56-0
MDL Number: MFCD09833624
Molecular Formula: C18H22N4O3
Molecular Weight: 342.40
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 106
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester

Formula: C₁₈H₂₂N₄O₃
MW: 342.40 g/mol
Melting Pt: 106 °C
MDL Number: MFCD09833624
CAS Number: 212322-56-0
https://us.vwr.com/stibo/web/std.lang.all/83/92/28968392.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR