2-Ethyl-1-hexanethiol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Ethylhexyl Mercaptan

E1031-100G E1031-25G
TCE1031-25GEA 60.27 USD
TCE1031-25G TCE1031-100G
2-Ethyl-1-hexanethiol ≥98.0% (by GC)
2-Ethyl-1-hexanethiol

CAS Number: 7341-17-5
MDL Number: MFCD00039654
Molecular Formula: C8H18S
Molecular Weight: 146.29
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Melting point (°C): -40
Flash Point (°C): 60
Specific Gravity (20/20): 0.85


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 2-Ethylhexyl Mercaptan

Formula: C₈H₁₈S
MW: 146.29 g/mol
Boiling Pt: 67 °C
Melting Pt: –40 °C
Density: 0.85 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00039654
CAS Number: 7341-17-5
https://us.vwr.com/stibo/web/std.lang.all/83/90/28968390.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR