Ethyl-3-Amino-4-methylbenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Amino-4-methylbenzoic Acid Ethyl Ester
, Ethyl 3-Amino-p-toluate
, 3-Amino-p-toluic Acid Ethyl Ester
, Ethyl 3-amino-4-methylbenzoate
CAS Number: 41191-92-8
MDL Number: MFCD01926538
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 51
Other Product Names / Synonyms: 3-Amino-4-methylbenzoic Acid Ethyl Ester, Ethyl 3-Amino-p-toluate, 3-Amino-p-toluic Acid Ethyl Ester
Formula:
C₁₀H₁₃NO₂ MW: 179.22 g/mol Boiling Pt: 105 °C Melting Pt: 51 °C |
MDL Number:
MFCD01926538 CAS Number: 41191-92-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...