1,1,2,3,3,3-Hexafluoropropyl ethyl ether ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 1,1,2,3,3,3-Hexafluoropropyl ether

E1020-25G E1020-5G
TCE1020-5GEA 127.65 USD
TCE1020-5G TCE1020-25G
1,1,2,3,3,3-Hexafluoropropyl ethyl ether ≥98.0% (by GC)
1,1,2,3,3,3-Hexafluoropropyl ethyl ether

CAS Number: 380-34-7
MDL Number: MFCD00236122
Molecular Formula: C5H6F6O
Molecular Weight: 196.09
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Specific Gravity (20/20): 1.32

Formula: C₅H₆F₆O
Boiling Pt: 65 °C
Density: 1.32 (20 °C)
MDL Number: MFCD00236122
CAS Number: 380-34-7
https://us.vwr.com/stibo/web/std.lang.all/83/80/28968380.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR