Ethyl-1,1,2,2-tetrafluoroethyl ether ≥98.0% (by GC)

Supplier: TCI America

E1019-25G E1019-5G
TCE1019-5GEA 18.27 USD
TCE1019-5G TCE1019-25G
Ethyl-1,1,2,2-tetrafluoroethyl ether ≥98.0% (by GC)
Ethyl-1,1,2,2-tetrafluoroethyl ether

CAS Number: 512-51-6
MDL Number: MFCD00069162
Molecular Formula: C4H6F4O
Molecular Weight: 146.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Flash Point (°C): -15
Specific Gravity (20/20): 1.21

Formula: C₄H₆F₄O
MW: 146.08 g/mol
Boiling Pt: 58 °C
Density: 1.21 (20 °C)
Flash Pt: –15 °C
MDL Number: MFCD00069162
CAS Number: 512-51-6
https://us.vwr.com/stibo/web/std.lang.all/83/79/28968379.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR