Ethyl-3-(trifluoromethyl)phenoxyacetate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 3-(Trifluoromethyl)phenoxyacetic Acid Ethyl Ester

E1004-1G
TCE1004-1GEA 555 USD
TCE1004-1G
Ethyl-3-(trifluoromethyl)phenoxyacetate ≥95.0% (by GC)
Ethyl-3-(trifluoromethyl)phenoxyacetate

CAS Number: 22897-99-0
MDL Number: MFCD03943910
Molecular Formula: C11H11F3O3
Molecular Weight: 248.20
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Specific Gravity (20/20): 1.26


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Trifluoromethyl)phenoxyacetic Acid Ethyl Ester

Formula: C₁₁H₁₁F₃O₃
MW: 248.20 g/mol
Boiling Pt: 123 °C
Density: 1.26 (20 °C)
MDL Number: MFCD03943910
CAS Number: 22897-99-0
https://us.vwr.com/stibo/web/std.lang.all/83/68/28968368.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR