Ethyl-1,3-dimethylpyrazole-5-carboxylate ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
Ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate
, 1,3-Dimethylpyrazole-5-carboxylic Acid Ethyl Ester
E1001-1G
TCE1001-1GEA
74
USD
TCE1001-1G
Ethyl-1,3-dimethylpyrazole-5-carboxylate ≥96.0% (by GC)
Ethyl-1,3-dimethylpyrazole-5-carboxylate
CAS Number: 5744-40-1
MDL Number: MFCD00233452
Molecular Formula: C8H12N2O2
Molecular Weight: 168.20
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 90
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 1,3-Dimethylpyrazole-5-carboxylic Acid Ethyl Ester
Formula:
C₈H₁₂N₂O₂ MW: 168.20 g/mol Boiling Pt: 90 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD00233452 CAS Number: 5744-40-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...