1-Ethyl-1-methylpyrrolidiniumtetrafluoroborate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
E0977-5G
E0977-25G
TCE0977-25GEA
276.08
USD
TCE0977-25G
TCE0977-5G
1-Ethyl-1-methylpyrrolidiniumtetrafluoroborate ≥98.0% (by total nitrogen basis)
1-Ethyl-1-methylpyrrolidinium tetrafluoroborate
CAS Number: 117947-85-0
MDL Number: MFCD08458505
Molecular Formula: C7H16BF4N
Molecular Weight: 201.02
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 286
Formula:
C₇H₁₆BF₄N Melting Pt: 286 °C |
MDL Number:
MFCD08458505 CAS Number: 117947-85-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...