(S)-Citalopram oxalate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate
CAS Number: 219861-08-2
MDL Number: MFCD06407826
Molecular Formula: C20H21FN2O
Molecular Weight: 414.43
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 13 deg (C=1, MeOH)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate
Formula:
C₂₀H₂₁FN₂O MW: 414.43 g/mol Melting Pt: 146 °C |
MDL Number:
MFCD06407826 CAS Number: 219861-08-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...