Ethyl cyclobutanecarboxylate ≥98.0% (by GC)

Supplier: TCI America

E0931-5G E0931-25G
TCE0931-5GEA 31 USD
TCE0931-5G TCE0931-25G
Ethyl cyclobutanecarboxylate ≥98.0% (by GC)
Ethyl cyclobutanecarboxylate

CAS Number: 14924-53-9
MDL Number: MFCD00001321
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 159
Flash Point (°C): 41
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Cyclobutanecarboxylic Acid Ethyl Ester

https://us.vwr.com/stibo/web/std.lang.all/83/10/28968310.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR