2-Ethynylthiophene ≥96.0% (by GC)
Supplier: TCI America
CAS Number: 4298-52-6
MDL Number: MFCD03425129
Molecular Formula: C6H4S
Molecular Weight: 108.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 46
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: (2-Thienyl)acetylene
Formula:
C₆H₄S MW: 108.1610 g/mol Boiling Pt: 46 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD03425129 CAS Number: 4298-52-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...