2-Ethyl-2-methacryloyloxyadamantane ≥97.0% (by GC) stabilized

Supplier: TCI America

Synonyms: 2-Ethyl-2-adamantyl MethacrylateMethacrylic Acid 2-Ethyl-2-adamantyl Ester

E0909-5G
TCE0909-5GEA 167.4 USD
TCE0909-5G
2-Ethyl-2-methacryloyloxyadamantane ≥97.0% (by GC) stabilized
2-Ethyl-2-methacryloyloxyadamantane

(stabilized with MEHQ)
CAS Number: 209982-56-9
MDL Number: MFCD06411144
Molecular Formula: C16H24O2
Molecular Weight: 248.37
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 40
Flash Point (°C): 144
Storage Temperature: 0°C


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: 2-Ethyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Ethyl-2-adamantyl Ester

Formula: C₁₆H₂₄O₂
MW: 248.37 g/mol
Melting Pt: 40 °C
Flash Pt: 144 °C
MDL Number: MFCD06411144
CAS Number: 209982-56-9
https://us.vwr.com/stibo/web/std.lang.all/82/92/28968292.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR