Eplerenone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester

E0905-200MG
TCE0905-200MGEA 156 USD
TCE0905-200MG
Eplerenone ≥98.0% (by HPLC)
Eplerenone

CAS Number: 107724-20-9
MDL Number: MFCD07783647
Molecular Formula: C24H30O6
Molecular Weight: 414.50
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 242
Specific rotation [a]20/D: 1.2 deg (C=1, CHCl3)
Lambda max.: 240 nmStorage Temperature: 0-10°C


Product Categories: Bioscience, Antihypertensive Ingredients, Other Steroids
Other Product Names / Synonyms: 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester

Formula: C₂₄H₃₀O₆
MW: 414.50 g/mol
Melting Pt: 242 °C
MDL Number: MFCD07783647
CAS Number: 107724-20-9
https://us.vwr.com/stibo/web/std.lang.all/82/89/28968289.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR