3-Ethynylthiophene ≥97.0% (by GC)

Supplier: TCI America

E0892-5G E0892-1G
TCE0892-5GEA 181 USD
TCE0892-5G TCE0892-1G
3-Ethynylthiophene ≥97.0% (by GC)
3-Ethynylthiophene

CAS Number: 67237-53-0
MDL Number: MFCD04039973
Molecular Formula: C6H4S
Molecular Weight: 108.16
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): 46
Specific Gravity (20/20): 1.09
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Thienylacetylene

Formula: C₆H₄S
MW: 108.16 g/mol
Boiling Pt: 50 °C
Density: 1.09 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD04039973
CAS Number: 67237-53-0
https://us.vwr.com/stibo/web/std.lang.all/82/76/28968276.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR