2-Ethoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0870-25G E0870-5G
TCE0870-25GEA 55.83 USD
TCE0870-25G TCE0870-5G
2-Ethoxypyridine ≥98.0% (by GC, titration analysis)
2-Ethoxypyridine

CAS Number: 14529-53-4
MDL Number: MFCD00023464
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 52
Specific Gravity (20/20): 1.01

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 157 °C
Density: 1.01 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00023464
CAS Number: 14529-53-4
https://us.vwr.com/stibo/web/std.lang.all/82/57/28968257.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR