Ethyl-1-benzyl-4-piperidinecarboxylate ≥95.0% (by GC, titration analysis)

Synonyms: 1-Benzyl-4-piperidinecarboxylic Acid Ethyl EsterEthyl 1-Benzylisonipecotate1-Benzylisonipecotic Acid Ethyl Ester

E0862-25G E0862-5G
TCE0862-25GEA 225.33 USD
TCE0862-25G TCE0862-5G
Ethyl-1-benzyl-4-piperidinecarboxylate ≥95.0% (by GC, titration analysis)
Ethyl-1-benzyl-4-piperidinecarboxylate

CAS Number: 24228-40-8
MDL Number: MFCD00040748
Molecular Formula: C15H21NO2
Molecular Weight: 247.34
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 136
Specific Gravity (20/20): 1.04


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: 1-Benzyl-4-piperidinecarboxylic Acid Ethyl Ester, Ethyl 1-Benzylisonipecotate, 1-Benzylisonipecotic Acid Ethyl Ester

Formula: C₁₅H₂₁NO₂
MW: 247.34 g/mol
Boiling Pt: 136 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00040748
CAS Number: 24228-40-8
https://us.vwr.com/stibo/web/std.lang.all/82/51/28968251.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR