Ethyl-3-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Fluorobenzoic acid ethyl ester

E0861-25G
TCE0861-25GEA 129.17 USD
TCE0861-25G
Ethyl-3-fluorobenzoate ≥98.0% (by GC)
Ethyl 3-fluorobenzoate

CAS Number: 451-02-5
MDL Number: MFCD00000336
Molecular Formula: C9H9FO2
Molecular Weight: 168.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 209
Melting point (°C): -34
Flash Point (°C): 82
Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluorobenzoic Acid Ethyl Ester

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 209 °C
Melting Pt: –34 °C
Density: 1.14 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00000336
CAS Number: 451-02-5
https://us.vwr.com/stibo/web/std.lang.all/82/50/28968250.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR