Etodolac ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Etodlic Acid1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic Acid

E0858-25G E0858-5G
TCE0858-5GEA 123.83 USD
TCE0858-5G TCE0858-25G
Etodolac ≥98.0% (by HPLC, titration analysis)
Etodolac

CAS Number: 41340-25-4
MDL Number: MFCD00133313
Molecular Formula: C17H21NO3
Molecular Weight: 287.36
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 148
Storage Temperature: 0-10°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Etodlic Acid, 1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic Acid

Formula: C₁₇H₂₁NO₃
MW: 287.36 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00133313
CAS Number: 41340-25-4
https://us.vwr.com/stibo/web/std.lang.all/82/47/28968247.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR