Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 4,5,6,7-Tetrahydroisoindole-1-carboxylic Acid Ethyl Ester

E0857-1G
TCE0857-1GEA 228.27 USD
TCE0857-1G
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate ≥95.0% (by GC)
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate

CAS Number: 65880-17-3
MDL Number: MFCD06227773
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 79
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4,5,6,7-Tetrahydroisoindole-1-carboxylic Acid Ethyl Ester

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Melting Pt: 79 °C
MDL Number: MFCD06227773
CAS Number: 65880-17-3
https://us.vwr.com/stibo/web/std.lang.all/82/46/28968246.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR