Ethylmagnesium chloride ∼1.0 mol/L in THF, activated with zinc chloride (ca. 10mol%)
Supplier: TCI America
E0778-250G
TCE0778-250GEA
114
USD
TCE0778-250G
Ethylmagnesium chloride ∼1.0 mol/L in THF, activated with zinc chloride (ca. 10mol%)
Ethylmagnesium chloride
CAS Number: 2386-64-3
Molecular Formula: C2H5ClMg
Molecular Weight: 88.82
Flash Point (°C): -17
Formula:
C₂H₅ClMg Flash Pt: –17 °C |
MDL Number:
MFCD00000472 CAS Number: 2386-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...