Ethyl-4-pentenoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl pent-4-enoate4-Pentenoic Acid Ethyl Ester

E0715-5G
TCE0715-5GEA 96.8 USD
TCE0715-5G
Ethyl-4-pentenoate ≥98.0% (by GC)
Ethyl-4-pentenoate

CAS Number: 1968-40-7
MDL Number: MFCD00036562
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 40
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: 4-Pentenoic Acid Ethyl Ester

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 146 °C
Density: 0.90 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00036562
CAS Number: 1968-40-7
https://us.vwr.com/stibo/web/std.lang.all/81/31/28968131.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR