S-Ethyl thioacetate ≥98.0% (by GC)
Supplier: TCI America
E0708-25G
TCE0708-25GEA
65.93
USD
TCE0708-25G
S-Ethyl thioacetate ≥98.0% (by GC)
S-Ethyl thioacetate
CAS Number: 625-60-5
MDL Number: MFCD00015178
Molecular Formula: C4H8OS
Molecular Weight: 104.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 18
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: Thioacetic Acid S-Ethyl Ester
Formula:
C₄H₈OS MW: 104.17 g/mol Boiling Pt: 117 °C Density: 0.98 (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00015178 CAS Number: 625-60-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...