trans-4-(4-Ethylcyclohexyl)benzonitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(trans-4-Ethylcyclohexyl)benzonitrile
E0703-5G
TCE0703-5GEA
69
USD
TCE0703-5G
trans-4-(4-Ethylcyclohexyl)benzonitrile ≥98.0% (by GC)
trans-4-(4-Ethylcyclohexyl)benzonitrile
CAS Number: 72928-54-2
Molecular Formula: C15H19N
Molecular Weight: 213.32
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 41
Other Product Names / Synonyms: 1-Cyano-4-(trans-4-ethylcyclohexyl)benzene
Formula:
C₁₅H₁₉N MW: 213.32 g/mol Melting Pt: 41 °C |
MDL Number:
MFCD09750932 CAS Number: 72928-54-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...