2-Ethoxypyrazine ≥98.0% (by GC)
Supplier: TCI America
E0697-5G
E0697-25G
TCE0697-5GEA
26.38
USD
TCE0697-5G
TCE0697-25G
2-Ethoxypyrazine ≥98.0% (by GC)
2-Ethoxypyrazine
CAS Number: 38028-67-0
MDL Number: MFCD00014609
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 92
Specific Gravity (20/20): 1.07
Formula:
C₆H₈N₂O MW: 125.15 g/mol Boiling Pt: 92 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD00014609 CAS Number: 38028-67-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...