1-Fluoro-2-ethynylbenzene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
(2-Fluorophenyl)acetylene
CAS Number: 766-49-4
Molecular Formula: C8H5F
Molecular Weight: 120.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 40
Flash Point (°C): 37
Specific Gravity (20/20): 1.06
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 2-Fluorophenylacetylene
Formula:
C₈H₅F MW: 120.13 g/mol Boiling Pt: 40 °C Density: 1.06 (20 °C) Flash Pt: 37 °C |
MDL Number:
MFCD00799540 CAS Number: 766-49-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...