all cis-8,11,14-Eicosatrienoic acid ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
all cis-8,11,14-Icosatrienoic Acid
CAS Number: 1783-84-2
MDL Number: MFCD00065721
Molecular Formula: C20H34O2
Molecular Weight: 306.49
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 62
Storage Temperature:
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols, Anti-inflammatory Ingredients
Other Product Names / Synonyms: all cis-8,11,14-Icosatrienoic Acid, Dihomo-gamma-linolenic Acid
Formula:
C₂₀H₃₄O₂ MW: 306.49 g/mol Boiling Pt: 200 °C Flash Pt: 62 °C |
MDL Number:
MFCD00065721 CAS Number: 1783-84-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...