1-Ethyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)

E0551-5G
TCE0551-5GEA 124.63 USD
TCE0551-5G
1-Ethyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)
1-Ethyl-1-phenylhydrazine

CAS Number: 644-21-3
MDL Number: MFCD00031413
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 119
Specific Gravity (20/20): 1.02

Formula: C₈H₁₂N₂
Boiling Pt: 119 °C
Density: 1,02 (20 °C)
MDL Number: MFCD00031413
CAS Number: 644-21-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR