1-Ethyl-3-methylimidazoliumtetrafluoroborate ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate
E0496-25G
E0496-5G
TCE0496-25GEA
221.14
USD
TCE0496-25G
TCE0496-5G
1-Ethyl-3-methylimidazoliumtetrafluoroborate ≥97.0% (by total nitrogen basis)
1-Ethyl-3-methylimidazolium tetrafluoroborate
[for Molten Salt]
CAS Number: 143314-16-3
MDL Number: MFCD00216668
Molecular Formula: C6H11BF4N2
Molecular Weight: 197.97
Purity/Analysis Method:
Form: Clear Liquid
Melting point (°C): 15
Flash Point (°C): 110
Specific Gravity (20/20): 1.29
Formula:
C₆H₁₁BF₄N₂ MW: 197.97 g/mol Melting Pt: 15 °C Density: 1,29 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00216668 CAS Number: 143314-16-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...