Ethyl malonyl chloride ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Ethyl Malonyl Chloride
CAS Number: 36239-09-5
MDL Number: MFCD00000736
Molecular Formula: C5H7ClO3
Molecular Weight: 150.56
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): 64
Specific Gravity (20/20): 1.19
Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: (Chloroformyl)acetic Acid Ethyl Ester, Ethyl (Chloroformyl)acetate, Ethyl 3-Chloro-3-oxopropionate, 3-Chloro-3-oxopropionic Acid Ethyl Ester
Formula:
C₅H₇ClO₃ MW: 150.56 g/mol Boiling Pt: 83 °C Density: 1.19 (20 °C) Flash Pt: 64 °C |
MDL Number:
MFCD00000736 CAS Number: 36239-09-5 UN: 3265 ADR: 8,II |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...