2-Ethylfuran ≥98.0% (by GC)

Supplier: TCI America

E0454-10ML
TCE0454-10MLEA 68.01 USD
TCE0454-10ML
2-Ethylfuran ≥98.0% (by GC)
2-Ethylfuran

CAS Number: 3208-16-0
MDL Number: MFCD00003259
Molecular Formula: C6H8O
Molecular Weight: 96.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): -2
Specific Gravity (20/20): 0.91

Formula: C₆H₈O
Boiling Pt: 92 °C
Density: 0.91 (20 °C)
Flash Pt: –2 °C
MDL Number: MFCD00003259
CAS Number: 3208-16-0
UN: 3271
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/79/69/28967969.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR