Ethyl isothiocyanatoformate ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
Ethoxycarbonyl isothiocyanate
E0440-25G
E0440-5G
TCE0440-5GEA
30
USD
TCE0440-5G
TCE0440-25G
Ethyl isothiocyanatoformate ≥95.0% (by GC)
Ethyl isothiocyanatoformate
CAS Number: 16182-04-0
MDL Number: MFCD00004814
Molecular Formula: C4H5NO2S
Molecular Weight: 131.15
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 50
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Ethyl Isothiocyanatoformate, Isothiocyanatoformic Acid Ethyl Ester
Formula:
C₄H₅NO₂S MW: 131.16 g/mol Boiling Pt: 61 °C Density: 1.16 (20 °C) Flash Pt: 50 °C |
MDL Number:
MFCD00004814 CAS Number: 16182-04-0 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...