2-Ethylthiophene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: o-Ethylthiophene

E0380-25ML
TCE0380-25MLEA 142 USD
TCE0380-25ML
2-Ethylthiophene ≥97.0% (by GC)
2-Ethylthiophene

CAS Number: 872-55-9
MDL Number: MFCD00005461
Molecular Formula: C6H8S
Molecular Weight: 112.19
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 21
Specific Gravity (20/20): 0.99

Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 137 °C
Density: 0.99 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00005461
CAS Number: 872-55-9
UN: 1993
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/79/03/28967903.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR