2-Ethylpyrazine ≥99.0% (by GC)

Supplier: TCI America

Synonyms: Ethylpyrazine

E0334-5G
TCE0334-5GEA 30.45 USD
TCE0334-5G
2-Ethylpyrazine ≥99.0% (by GC)
2-Ethylpyrazine

CAS Number: 13925-00-3
MDL Number: MFCD00006149
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 42
Specific Gravity (20/20): 1.00

Formula: C₆H₈N₂
Boiling Pt: 153 °C
Density: 1.00 (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD00006149
CAS Number: 13925-00-3
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/78/65/28967865.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR