2-Ethyl-2-imidazoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E0234-5G
TCE0234-5GEA
108.57
USD
TCE0234-5G
2-Ethyl-2-imidazoline ≥98.0% (by GC, titration analysis)
2-Ethyl-2-imidazoline
CAS Number: 930-52-9
MDL Number: MFCD00242607
Molecular Formula: C5H10N2
Molecular Weight: 98.15
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 44
Formula:
C₅H₁₀N₂ MW: 98.15 g/mol Boiling Pt: 120 °C Melting Pt: 44 °C |
MDL Number:
MFCD00242607 CAS Number: 930-52-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...