1-Ethylquinaldinium iodide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Ethyl-2-methylquinolin-1-ium iodide1-Ethyl-2-methylquinolinium iodide

E0172-25G
TCE0172-25GEA 140.83 USD
TCE0172-25G
1-Ethylquinaldinium iodide ≥99.0% (by titrimetric analysis)
1-Ethylquinaldinium iodide

CAS Number: 606-55-3
MDL Number: MFCD00031829
Molecular Formula: C12H14IN
Molecular Weight: 299.16
Purity/Analysis Method: 99.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1-Ethylquinaldinium Iodide

Formula: C₁₂H₁₄IN
MW: 299.15 g/mol
MDL Number: MFCD00031829
CAS Number: 606-55-3
https://us.vwr.com/stibo/web/std.lang.all/77/47/28967747.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR