3-Ethylpyridine ≥98.0% (by GC)

Supplier: TCI America

E0169-25ML
TCE0169-25MLEA 83.23 USD
TCE0169-25ML
3-Ethylpyridine ≥98.0% (by GC)
3-Ethylpyridine

CAS Number: 536-78-7
MDL Number: MFCD00006413
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 48
Specific Gravity (20/20): 0.94

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 165 °C
Density: 0.94 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00006413
CAS Number: 536-78-7
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/77/44/28967744.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR