Ethanesulfonic acid ≥90.0% (by titrimetric analysis)

Supplier: TCI America

E0033-25G
TCE0033-25GEA 111.65 USD
TCE0033-25G
Ethanesulfonic acid ≥90.0% (by titrimetric analysis)
Ethanesulphonic acid

CAS Number: 594-45-6
MDL Number: MFCD00007529
Molecular Formula: C2H6O3S
Molecular Weight: 110.13
Purity/Analysis Method: 90.0% (T)
Form: Clear Liquid
Boiling point (°C): 140
Melting point (°C): -17
Flash Point (°C): 110
Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: Ethylsulfonic Acid

Formula: C₂H₆O₃S
MW: 110.13 g/mol
Boiling Pt: 140 °C
Melting Pt: –17 °C
Density: 1.34 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00007529
CAS Number: 594-45-6
UN: 2586
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/76/39/28967639.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR