Dipropargylamine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Di(2-propynyl)amine

D4685-5ML D4685-1ML
TCD4685-5MLEA 373.6 USD
TCD4685-5ML TCD4685-1ML
Dipropargylamine ≥97.0% (by titrimetric analysis)
Dipropargylamine

CAS Number: 6921-28-4
MDL Number: MFCD00008574
Molecular Formula: C6H7N
Molecular Weight: 93.13
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Boiling point (°C): 65
Flash Point (°C): 46
Specific Gravity (20/20): 0.91


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Di(2-propynyl)amine

Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 65 °C
Density: 0.91 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00008574
CAS Number: 6921-28-4
https://us.vwr.com/stibo/web/std.lang.all/68/20/28986820.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR