Diethylene glycol isopropyl methyl ether ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-[2-(2-Methoxyethoxy)ethoxy]propane

D4677-500ML D4677-25ML
TCD4677-500MLEA 112.62 USD
TCD4677-500ML TCD4677-25ML
Diethylene glycol isopropyl methyl ether ≥98.0% (by GC)
Diethylene glycol isopropyl methyl ether

CAS Number: 77078-22-9
MDL Number: MFCD28009631
Molecular Formula: C8H18O3
Molecular Weight: 162.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 2-[2-(2-Methoxyethoxy)ethoxy]propane

Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Density: 0.91 (20 °C)
MDL Number: MFCD28009631
CAS Number: 77078-22-9
https://us.vwr.com/stibo/web/std.lang.all/68/12/28986812.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR