1,5-Dimethyl-1H-pyrazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D4630-5ML
TCD4630-5MLEA 128 USD
TCD4630-5ML
1,5-Dimethyl-1H-pyrazole ≥98.0% (by titrimetric analysis)
1,5-Dimethyl-1H-pyrazole

CAS Number: 694-31-5
MDL Number: MFCD01646129
Molecular Formula: C5H8N2
Molecular Weight: 96.13
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 0.98
Lambda max.: 217 nm (EtOH)

Formula: C₅H₈N₂
MW: 96.1300 g/mol
Boiling Pt: 158 °C
Density: 0.98 (20 °C)
MDL Number: MFCD01646129
CAS Number: 694-31-5
https://us.vwr.com/stibo/web/std.lang.all/67/71/28986771.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR