9,9-Di(p-tolyl)fluorene ≥98.0%
Supplier: TCI America
D4610-1G
D4610-5G
TCD4610-1GEA
101
USD
TCD4610-1G
TCD4610-5G
9,9-Di(p-tolyl)fluorene ≥98.0%
9,9-Di(p-tolyl)fluorene
CAS Number: 54941-50-3
MDL Number: MFCD23115707
Molecular Formula: C27H22
Molecular Weight: 346.47
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 169
Lambda max.: 305 nm (CH2Cl2)
Formula:
C₂₇H₂₂ Melting Pt: 169 °C |
MDL Number:
MFCD23115707 CAS Number: 54941-50-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...