TCIAD4525

D4525-5G D4525-1G
TCD4525-5GEA 336.21 USD
TCD4525-5G TCD4525-1G
TCIAD4525
CAS Number: 94-98-4
MDL Number: MFCD00025575
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 219
Specific Gravity (20/20): 0.96
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR