2,4-Dimethylbenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Hydroxymethyl)-m-xylene
, (2,4-Dimethylphenyl)methanol
D4519-5G
D4519-25G
TCD4519-5GEA
140.68
USD
TCD4519-5G
TCD4519-25G
2,4-Dimethylbenzyl alcohol ≥98.0% (by GC)
2,4-Dimethylbenzyl alcohol
CAS Number: 16308-92-2
MDL Number: MFCD00004624
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 120
Flash Point (°C): 113
Freezing point (°C): 25
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 120 °C Flash Pt: 113 °C |
MDL Number:
MFCD00004624 CAS Number: 16308-92-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...