Diethyl-2,2':6',2''-terpyridine-4'-phosphonate ≥93.0% (by GC)

Supplier: TCI America

Synonyms: 2,2':6',2''-Terpyridine-4'-phosphonic Acid Diethyl Ester

D4511-200MG
TCD4511-200MGEA 432 USD
TCD4511-200MG
Diethyl-2,2':6',2''-terpyridine-4'-phosphonate ≥93.0% (by GC)
Diethyl-2,2':6',2''-terpyridine-4'-phosphonate

CAS Number: 161583-75-1
MDL Number: MFCD11042551
Molecular Formula: C19H20N3O3P
Molecular Weight: 369.36
Purity/Analysis Method: 93.0% (GC)
Form: Crystal
Melting point (°C): 149


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 2,2':6',2''-Terpyridine-4'-phosphonic Acid Diethyl Ester

Formula: C₁₉H₂₀N₃O₃P
MW: 369.36 g/mol
Melting Pt: 149 °C
MDL Number: MFCD11042551
CAS Number: 161583-75-1
https://us.vwr.com/stibo/web/std.lang.all/66/38/28986638.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR