5,8-Dibromo-2,3-diphenylquinoxaline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4461-200MG
TCD4461-200MGEA 196 USD
TCD4461-200MG
5,8-Dibromo-2,3-diphenylquinoxaline ≥98.0% (by HPLC, total nitrogen)
5,8-Dibromo-2,3-diphenylquinoxaline

CAS Number: 94544-77-1
Molecular Formula: C20H12Br2N2
Molecular Weight: 440.14
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 222
Lambda max.: 358 nm (CHCl3)

Formula: C₂₀H₁₂Br₂N₂
MW: 440.14 g/mol
Melting Pt: 222 °C
MDL Number: MFCD27966265
CAS Number: 94544-77-1
https://us.vwr.com/stibo/web/std.lang.all/65/84/28986584.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR