3,5-Dichloropyridazine ≥98.0% (by GC)

Supplier: TCI America

D4447-1G
TCD4447-1GEA 72.05 USD
TCD4447-1G
3,5-Dichloropyridazine ≥98.0% (by GC)
3,5-Dichloropyridazine

CAS Number: 1837-55-4
MDL Number: MFCD10698048
Molecular Formula: C4H2Cl2N2
Molecular Weight: 148.97
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 62
Storage Temperature: 0-10°C

Formula: C₄H₂Cl₂N₂
MW: 148.9781 g/mol
Melting Pt: 62 °C
MDL Number: MFCD10698048
CAS Number: 1837-55-4
https://us.vwr.com/stibo/web/std.lang.all/65/70/28986570.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR