D 358 ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
12-[3-(Carboxymethyl)-5-[[4-[4-(2,2-diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]methylene]-4,4'-dioxo-2'-thioxo-4,5-dihydro-2'H,3H-[2,5'-bithiazolylidene]-3'(4'H)-yl]dodecanoic Acid
CAS Number: 1207638-53-6
Molecular Formula: C52H53N3O6S3
Molecular Weight: 912.19
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Solar Cell Research Reagents
Other Product Names / Synonyms: 12-[3-(Carboxymethyl)-5-[[4-[4-(2,2-diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]methylene]-4,4'-dioxo-2'-thioxo-4,5-dihydro-2'H,3H-[2,5'-bithiazolylidene]-3'(4'H)-yl]dodecanoic Acid
Formula:
C₅₂H₅₃N₃O₆S₃ MW: 912.19 g/mol |
CAS Number:
1207638-53-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...