D 102 ≥96.0% (by HPLC)
Supplier: TCI America
Synonyms:
2-[5-[[4-[4-(2,2-Diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]methylene]-4-oxo-2-thioxothiazolidin-3-yl]acetic Acid
CAS Number: 652145-28-3
Molecular Formula: C37H30N2O3S2
Molecular Weight: 614.78
Purity/Analysis Method:
Form: Crystal
Lambda max.: 505 nm (CH2Cl2)
Product Categories: Materials Chemistry, Solar Cell Research Reagents
Other Product Names / Synonyms: 2-[5-[[4-[4-(2,2-Diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]methylene]-4-oxo-2-thioxothiazolidin-3-yl]acetic Acid
Formula:
C₃₇H₃₀N₂O₃S₂ MW: 614.78 g/mol |
CAS Number:
652145-28-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...