1,3,2-Dioxathiane-2-oxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Propylene SulfiteTrimethylene Sulfite1,3-Propanediol Cyclic Sulfite

D4426-25G
TCD4426-25GEA 51.75 USD
TCD4426-25G
1,3,2-Dioxathiane-2-oxide ≥98.0% (by GC)
1,3,2-Dioxathiane-2-oxide

CAS Number: 4176-55-0
MDL Number: MFCD01682880
Molecular Formula: C3H6O3S
Molecular Weight: 122.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 54
Flash Point (°C): 84
Freezing point (°C): -14
Specific Gravity (20/20): 1.35


Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis)
Other Product Names / Synonyms: 1,3-Propylene Sulfite, Trimethylene Sulfite, 1,3-Propanediol Cyclic Sulfite

Formula: C₃H₆O₃S
MW: 122.14 g/mol
Boiling Pt: 54 °C
Density: 1.35 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD01682880
CAS Number: 4176-55-0
https://us.vwr.com/stibo/web/std.lang.all/65/37/28986537.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR