Dimethyl-4-nitrophthalate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Dimethyl 4-nitrophthalate
D4424-1G
D4424-5G
TCD4424-5GEA
112
USD
TCD4424-5G
TCD4424-1G
Dimethyl-4-nitrophthalate ≥98.0% (by GC)
Dimethyl 4-nitrophthalate
CAS Number: 610-22-0
MDL Number: MFCD00017191
Molecular Formula: C10H9NO6
Molecular Weight: 239.18
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 67
Other Product Names / Synonyms: 4-Nitrophthalic Acid Dimethyl Ester
Formula:
C₁₀H₉NO₆ MW: 239.19 g/mol Boiling Pt: 195 °C Melting Pt: 67 °C |
MDL Number:
MFCD00017191 CAS Number: 610-22-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...